missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl Acetate 98.0+%, TCI America™

Catalog No. P182125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P182125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P182125G Supplier TCI America Supplier No. P182125G

Chemical Identifiers

CAS 326-61-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00014574
InChI Key PFWYHTORQZAGCA-UHFFFAOYSA-N
Synonym Acetic Acid Piperonyl Ester, 3,4-Methylenedioxybenzyl Acetate
PubChem CID 9473
IUPAC Name 1,3-benzodioxol-5-ylmethyl acetate
SMILES CC(=O)OCC1=CC2=C(C=C1)OCO2

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Piperonyl Acetate
EINECSNumber (5)-0513
RTECSNumber AI9050000
TSCA Yes