missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperitone (mixture of enantiomers, predominantly (R)-(-)-form) 94.0+%, TCI America™
Supplier: TCI America P178325G
Specifications
| Piperitone (mixture of enantiomers, predominantly (R)-(-)-form) | |
| -29°C | |
| 235°C | |
| 25 g | |
| YSTPAHQEHQSRJD-UHFFFAOYSA-N | |
| 3-methyl-6-propan-2-ylcyclohex-2-en-1-one | |
| 6987 | |
| 152.24 | |
| Liquid |
| 89-81-6 | |
| Yellow | |
| C10H16O | |
| p-Menth-1-en-3-one, 6-Isopropyl-3-methyl-2-cyclohexen-1-one | |
| CC1=CC(=O)C(CC1)C(C)C | |
| 152.237 | |
| CHEBI:48933 | |
| ≥94.0% (GC) |
Chemical Identifiers
| 89-81-6 | |
| 152.237 | |
| p-Menth-1-en-3-one, 6-Isopropyl-3-methyl-2-cyclohexen-1-one | |
| CHEBI:48933 | |
| CC1=CC(=O)C(CC1)C(C)C |
| C10H16O | |
| YSTPAHQEHQSRJD-UHFFFAOYSA-N | |
| 6987 | |
| 3-methyl-6-propan-2-ylcyclohex-2-en-1-one |
Safety and Handling
EINECSNumber : (3)-2393
RTECSNumber : OT0257000
TSCA : Yes