missing translation for 'onlineSavingsMsg'
Learn More

Piperine 97.0+%, TCI America™

Catalog No. p-7134619
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P04601G 1 g
P04605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P04601G Supplier TCI America Supplier No. P04601G

Chemical Identifiers

CAS 94-62-2
Molecular Formula C17H19NO3
Molecular Weight (g/mol) 285.34
MDL Number MFCD00005839
InChI Key MXXWOMGUGJBKIW-YPCIICBESA-N
Synonym piperine, 1-piperoylpiperidine, piperin, bioperine, piperoylpiperidine, e,e-1-piperoylpiperidine, n-e,e-piperoyl piperidine, piperidine, 1-piperoyl-, e,e, unii-u71xl721qk, fema no. 2909
PubChem CID 638024
ChEBI CHEBI:28821
IUPAC Name (2E,4E)-5-(2H-1,3-benzodioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one
SMILES O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
Formula Weight 285.34
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Piperine
RTECSNumber TN2321500
TSCA Yes