missing translation for 'onlineSavingsMsg'
Learn More

Piperine, 98%

Catalog No. AAA1351006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1351006 5 g
AAA1351014 25 g
AAA1351022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1351006 Supplier Thermo Scientific Chemicals Supplier No. A1351006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94-62-2 | C17H19NO3 | 285.34 g/mol

Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

Solubility
Soluble in water(0.04g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 94-62-2
Molecular Formula C17H19NO3
Molecular Weight (g/mol) 285.34
MDL Number MFCD00005839
InChI Key MXXWOMGUGJBKIW-YPCIICBESA-N
Synonym piperine, 1-piperoylpiperidine, piperin, bioperine, piperoylpiperidine, e,e-1-piperoylpiperidine, n-e,e-piperoyl piperidine, piperidine, 1-piperoyl-, e,e, unii-u71xl721qk, fema no. 2909
PubChem CID 638024
ChEBI CHEBI:28821
IUPAC Name (2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
SMILES O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1

Specifications

Melting Point 128°C to 133°C
Quantity 5 g
Beilstein 90741
Merck Index 14,7472
Solubility Information Soluble in water(0.04g/L).
Formula Weight 285.34
Percent Purity 98%
Chemical Name or Material Piperine
Hazard Category H302
Hazard Statement GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-348-0
RTECSNumber TN2321500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only