missing translation for 'onlineSavingsMsg'
Learn More

Piperidine-1-sulfonyl chloride, 97%

Catalog No. p-7035834
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54053MD 250 mg
AAH5405303 1 g
AAH5405306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54053MD Supplier Thermo Scientific Chemicals Supplier No. H54053MD

CAS: 35856-62-3 | C5H10ClNO2S | 183.65 g/mol

Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 35856-62-3
Molecular Formula C5H10ClNO2S
Molecular Weight (g/mol) 183.65
MDL Number MFCD03250329
InChI Key QQJYAXDCMMXECR-UHFFFAOYSA-N
Synonym 1-piperidinesulfonyl chloride, chloropiperidylsulfone, piperidinsulfochlorid, piperidinesulfonyl chloride, piperidinosulfonyl chloride, pperdne-1-sulfonyl chlorde, 1-piperidinesulfonylchloride, piperidinyl sulfonyl chloride, 1-piperidine sulfonyl chloride, piperidine-1-sulfonic acid chloride
PubChem CID 11298344
IUPAC Name piperidine-1-sulfonyl chloride
SMILES C1CCN(CC1)S(=O)(=O)Cl

Specifications

Density 1.308
Refractive Index 1.492
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 183.65
Percent Purity 97%
Chemical Name or Material Piperidine-1-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only