missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Hexahydrate 98.0+%, TCI America™
$39.14 - $93.21
Chemical Identifiers
| CAS | 142-63-2 |
|---|---|
| Molecular Formula | C4H22N2O6 |
| Molecular Weight (g/mol) | 194.228 |
| MDL Number | MFCD00149389 |
| InChI Key | AVRVZRUEXIEGMP-UHFFFAOYSA-N |
| Synonym | piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn |
| PubChem CID | 120181 |
| IUPAC Name | piperazine;hexahydrate |
| SMILES | C1CNCCN1.O.O.O.O.O.O |
Chemical Identifiers
| 142-63-2 | |
| 194.228 | |
| AVRVZRUEXIEGMP-UHFFFAOYSA-N | |
| 120181 | |
| C1CNCCN1.O.O.O.O.O.O |
| C4H22N2O6 | |
| MFCD00149389 | |
| piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn | |
| piperazine;hexahydrate |
Specifications
| 142-63-2 | |
| White | |
| MFCD00149389 | |
| 2579 | |
| AVRVZRUEXIEGMP-UHFFFAOYSA-N | |
| piperazine;hexahydrate | |
| 120181 | |
| ≥98.0% (GC,T) | |
| Piperazine Hexahydrate |
| 44°C | |
| C4H22N2O6 | |
| 25 g | |
| piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn | |
| C1CNCCN1.O.O.O.O.O.O | |
| 194.228 | |
| 86.14 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0953
RTECSNumber : TM0850000
TSCA : No