missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Hexahydrate 98.0+%, TCI America™

Catalog No. P044725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044725G 25 g
P0447500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044725G Supplier TCI America Supplier No. P044725G

Chemical Identifiers

CAS 142-63-2
Molecular Formula C4H22N2O6
Molecular Weight (g/mol) 194.228
MDL Number MFCD00149389
InChI Key AVRVZRUEXIEGMP-UHFFFAOYSA-N
Synonym piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
PubChem CID 120181
IUPAC Name piperazine;hexahydrate
SMILES C1CNCCN1.O.O.O.O.O.O

Specifications

Melting Point 44°C
Color White
Quantity 25 g
UN Number 2579
Formula Weight 86.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Hexahydrate
EINECSNumber (5)-0953
RTECSNumber TM0850000
TSCA No