missing translation for 'onlineSavingsMsg'
Learn More

Piperazine hexahydrate, 98%

CAS: 142-63-2 | C4H10N2·6H2O | 194.23 g/mol

$66.60 - $132.17

Chemical Identifiers

CAS 142-63-2
Molecular Formula C4H10N2·6H2O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00149389
InChI Key AVRVZRUEXIEGMP-UHFFFAOYSA-N
Synonym piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
PubChem CID 120181
IUPAC Name piperazine;hexahydrate
SMILES C1CNCCN1.O.O.O.O.O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131301000
SDS View Documents
Thermo Scientific Chemicals
131301000
100 g Plastic bottle
Each for $66.60
Only null left
 
AC131305000
SDS View Documents
Thermo Scientific Chemicals
131305000
500 g Plastic bottle
Each for $132.17
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

142-63-2
194.23
AVRVZRUEXIEGMP-UHFFFAOYSA-N
120181
C1CNCCN1.O.O.O.O.O.O
C4H10N2·6H2O
MFCD00149389
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
piperazine;hexahydrate

Specifications

142-63-2
White
87°C
98%
C4H10N2·6H2O
100 g
14,7464
Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
C1CNCCN1.O.O.O.O.O.O
194.23
194.23
Adhering Crystals
41.0°C to 45.0°C
145.0°C to 156.0°C
Authentic
Plastic bottle
MFCD00149389
23,4
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
AVRVZRUEXIEGMP-UHFFFAOYSA-N
piperazine;hexahydrate
120181
98%
Piperazine hexahydrate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

RTECSNumber : TM0850000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only