missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Dihydrochloride 98.0+%, TCI America™

Catalog No. P24915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24915G 5 g
P24911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24915G Supplier TCI America Supplier No. P24915G
Only null left

Chemical Identifiers

CAS 142-64-3
Molecular Formula C4H12Cl2N2
Molecular Weight (g/mol) 159.054
MDL Number MFCD00012754
InChI Key CVVIJWRCGSYCMB-UHFFFAOYSA-N
Synonym piperazine dihydrochloride, dowzene dhc, piperazine, dihydrochloride, dihydro pip wormer, caswell no. 668, piperazine wormer premix, unii-17vu4z4w88, diethylenediamine dihydrochloride, piperazine hcl, epa pesticide chemical code 067401
PubChem CID 8893
IUPAC Name piperazine;dihydrochloride
SMILES C1CNCCN1.Cl.Cl

Specifications

Melting Point 320°C
Color White
Quantity 5 g
Formula Weight 159.05
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Dihydrochloride
RTECSNumber TL4025000
TSCA Yes