missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Dihydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 320°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 159.05 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Piperazine Dihydrochloride |
Safety and Handling
| RTECSNumber | TL4025000 |
| TSCA | Yes |