missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Dihydrobromide 98.0+%, TCI America™

Catalog No. p-7135583
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24901G 1 g
P24905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24901G Supplier TCI America Supplier No. P24901G
Only null left

Chemical Identifiers

CAS 59813-05-7
Molecular Formula C4H12Br2N2
Molecular Weight (g/mol) 247.962
InChI Key UXWKNNJFYZFNDI-UHFFFAOYSA-N
Synonym Piperazinium Dibromide, Piperazinium Bromide
PubChem CID 3017157
IUPAC Name piperazine;dihydrobromide
SMILES C1CNCCN1.Br.Br

Specifications

Melting Point 360°C
Color White
Quantity 1 g
Formula Weight 247.96
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Dihydrobromide