missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Citrate Hydrate 98.0+%, TCI America™

$57.79 - $132.35

Chemical Identifiers

CAS 144-29-6
Molecular Formula C4H10N2
MDL Number MFCD00038961
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P044925G
View Documents
TCI America
P044925G
25 g
Each for $57.79
Only null left
 
P0449500G
View Documents
TCI America
P0449500G
500 g
Each for $132.35
Only null left
 
Description

Description

Specifications

Chemical Identifiers

144-29-6
C4H10N2
MFCD00038961

Specifications

144-29-6
White
MFCD00038961
642.66
Crystalline Powder
185°C
C4H10N2
25 g
≥98.0% (T)
Piperazine Citrate Hydrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TL3485000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'