missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Citrate Hydrate 98.0+%, TCI America™

Catalog No. P0449500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0449500G 500 g
P044925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0449500G Supplier TCI America Supplier No. P0449500G
Only null left

Chemical Identifiers

CAS 144-29-6
Molecular Formula C4H10N2
MDL Number MFCD00038961

Specifications

Melting Point 185°C
Color White
Quantity 500 g
Formula Weight 642.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Citrate Hydrate
RTECSNumber TL3485000
TSCA Yes