missing translation for 'onlineSavingsMsg'
Learn More

Piperazine-1,4-bis(2-ethanesulfonic Acid) Monosodium Salt 99.0+%, TCI America™

Catalog No. P087525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087525G Supplier TCI America Supplier No. P087525G

Chemical Identifiers

CAS 10010-67-0
Molecular Formula C8H17N2NaO6S2
Molecular Weight (g/mol) 324.34
MDL Number MFCD00065472
InChI Key OGGAIRCLBMGXCZ-UHFFFAOYSA-M
Synonym pipes monosodium salt, pipes monosodium, sodium 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonate, 1,4-piperazinediethanesulfonic acid, monosodium salt, sodium pipes, pipes sodium salt, piperazine-1,4-diethanesulfonate, sodium salt, unii-4j0k2g9mcw, sodium hydrogen piperazine-1,4-diethanesulphonate, 1,4-piperazinediethanesulfonic acid, sodium salt 1:1
PubChem CID 23670851
ChEBI CHEBI:63077
IUPAC Name sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Color White
Quantity 25 g
Formula Weight 324.34
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine-1,4-bis(2-ethanesulfonic Acid) Monosodium Salt [Good′s buffer component for biologi