missing translation for 'onlineSavingsMsg'
Learn More

Piperazine-1,4-bis(2-ethanesulfonic Acid) Disodium Salt 98.0+%, TCI America™

Catalog No. P087425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087425G Supplier TCI America Supplier No. P087425G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 76836-02-7
Molecular Formula C8H16N2Na2O6S2
Molecular Weight (g/mol) 346.324
MDL Number MFCD00064349
InChI Key GMHSTJRPSVFLMT-UHFFFAOYSA-L
Synonym pipes disodium salt, 1,4-piperazinediethanesulfonic acid, disodium salt, disodium piperazine-1,4-diethanesulphonate, unii-o25162tu3u, sodium 2,2'-piperazine-1,4-diyl diethanesulfonate, piperazine-1,4-bis 2-ethanesulfonic acid disodium salt, disodium 2-4-2-sulfonatoethyl piperazin-1-yl ethanesulfonate, 1,4-piperazinediethanesulfonic acid disodium salt, 1,4-piperazinediethanesulfonic acid, sodium salt 1:2, piperazine, n,n'-bis 2-ethanesulfonic acid disodium salt
PubChem CID 173553
ChEBI CHEBI:63055
IUPAC Name disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 346.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine-1,4-bis(2-ethanesulfonic Acid) Disodium Salt [Good′s buffer component for biologica