missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pipamperone 98.0+%, TCI America™
$309.43 - $832.11
Chemical Identifiers
| CAS | 1893-33-0 |
|---|---|
| Molecular Formula | C21H30FN3O2 |
| Molecular Weight (g/mol) | 375.488 |
| MDL Number | MFCD00242979 |
| InChI Key | AXKPFOAXAHJUAG-UHFFFAOYSA-N |
| Synonym | Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide |
| PubChem CID | 4830 |
| ChEBI | CHEBI:78549 |
| IUPAC Name | 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide |
| SMILES | C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P231525MG
|
TCI America
P231525MG |
25 mg |
Each for $309.43
|
|
|||||
|
P2315100MG
|
TCI America
P2315100MG |
100 mg |
Each for $832.11
|
|
|||||
Chemical Identifiers
| 1893-33-0 | |
| 375.488 | |
| AXKPFOAXAHJUAG-UHFFFAOYSA-N | |
| 4830 | |
| 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide |
| C21H30FN3O2 | |
| MFCD00242979 | |
| Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide | |
| CHEBI:78549 | |
| C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N |
Specifications
| 1893-33-0 | |
| White-Yellow | |
| MFCD00242979 | |
| 2811 | |
| AXKPFOAXAHJUAG-UHFFFAOYSA-N | |
| 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide | |
| 4830 | |
| 375.49 | |
| Crystalline Powder |
| 126°C | |
| C21H30FN3O2 | |
| 25 mg | |
| Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide | |
| C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N | |
| 375.488 | |
| CHEBI:78549 | |
| ≥98.0% (HPLC) | |
| Pipamperone |
Safety and Handling
RTECSNumber : DW1050000
TSCA : No