missing translation for 'onlineSavingsMsg'
Learn More

Pipamperone 98.0+%, TCI America™

Catalog No. P231525MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P231525MG 25 mg
P2315100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P231525MG Supplier TCI America Supplier No. P231525MG
Only null left

Chemical Identifiers

CAS 1893-33-0
Molecular Formula C21H30FN3O2
Molecular Weight (g/mol) 375.488
MDL Number MFCD00242979
InChI Key AXKPFOAXAHJUAG-UHFFFAOYSA-N
Synonym Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide
PubChem CID 4830
ChEBI CHEBI:78549
IUPAC Name 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide
SMILES C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 375.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pipamperone
RTECSNumber DW1050000
TSCA No