missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PIP4K2A (Human) Recombinant Protein

Catalog No. 89339433
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89339433 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89339433 Supplier Abnova™ Supplier No. P6571
Only null left

For use with Functional Study.

Human PIP4K2A (NP_005019, 1 a.a. - 406 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.

Specifications

For Use With (Application) Functional Study
Gene ID (Entrez) 5305
Name phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
Quality Control Testing The purity was assessed by SDS-PAGE/CBB staining.
Quantity 5 μg
Storage Requirements Stored at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ13267, PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK