missing translation for 'onlineSavingsMsg'
Learn More

PIP™ CleanTeam™ Polyurethane-Coated Nylon Clean Environment Gloves

Catalog No. 296357195
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296357195 Large
296357193 Small
296357194 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296357195 Supplier PIP™ Supplier No. S13PU9

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seamless knit nylon shell is comfortable and reduces hand fatigue, while the polyurethane coating provides a secure grip in dry, or slightly wet conditions.

  • Knit wrist helps prevent dirt and debris from entering the glove
  • Constructed from nylon filament yarns for low lint applications
  • Nylon yarns absorb perspiration to leave hands cool and comfortable
  • Can be worn as a liner under disposable and other liquid-proof gloves
  • Color coded hems for easy size identification

Specifications

Coated Area Fingertips
Construction Seamless Knit
Coating Material Polyurethane
Color White
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Product Handling, Electronics Assembly, Photography Labs, and Computer Manufacturing
Glove Cuff Color Green
Glove Cuff Style Knit
Series 40-C125
Length (English) 9.4 in.
Length (Metric) 24 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov