missing translation for 'onlineSavingsMsg'
Learn More

PIP™ Assurance™ 52-3672 Unsupported Neoprene Coated Latex Gloves with Raised Diamond Grip

Catalog No. 19161646C
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19161646C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19161646C Supplier PIP™ Supplier No. GP523672L
Only null left

Double-dipped neoprene construction provides protection against a wide range of chemicals such as acids, caustics, solvents, and oils. Raised diamond pattern enhances grip capabilities in wet or oily conditions.

  • Neoprene coating provides protection from a wide range of chemicals such as acids, caustics, solvents, and oils
  • Double dipped construction combines the versatile chemical resistance properties of neoprene with the comfort of latex
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Flock lining increases comfort and eases donning/doffing
  • Raised diamond pattern enhances grip capabilities
  • All component materials comply with Federal regulations for food contact 21 CFR, 170-199

Specifications

Certifications/Compliance 21 CFR, 170-199
Coating Material Neoprene
Color Blue, Yellow
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material Latex
Product Type Dip-coated Gloves
Size Large
Thickness (English) 19 mil
EN388 Standard 1X11
For Use With (Application) General Maintenance, Janitorial Work, Petrochemical, and Photo Finishing
Glove Cuff Style Straight
Liner Material Flocked Cotton
Series 52-3672
Length (English) 12.6 in.
Length (Metric) 32 cm
Show More Show Less