missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ Pinnacle Portfolio Clarifier™ 1, 2
Unique acetic acid reagents, developed to supplement our family of Hematoxylins
$514.74 - $514.74
Specifications
| Color | Clear |
|---|---|
| Format | Liquid |
| For Use With (Application) | Developed to supplement our family of hematoxylins |
| Quantity | 4 x 1 gal. |
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
22-220-107
|
Fisher Healthcare™
220107 |
Clarifier 1 |
Case of 4 Each for $514.74
|
|
|||||
Description
- Designed as a system
- Removes background stain caused by excessive use of tissue adhesive
- Increases cell transparency
- Selectively removes hematoxylin staining caused by excess adhesive without affecting nuclear staining
- Use of Clarifier will improve contrast between the nuclei and cytoplasm
- Acid alcohol requires quick dips while Clarifier dictates a set time (30-60 seconds for Clarifier 1 and 20-30 seconds for Clarifier 2)
- Each component has added value and benefits that reduces cost per slide
- Unique chemical formulae promotes long-term chemical stability, which extends the usable life of each component
- Subjected to some of the highest quality manufacturing standards in the industry
- Packaging improvements enhance product longevity
Specifications
| Clear | |
| Liquid | |
| Developed to supplement our family of hematoxylins | |
| 4 x 1 gal. |