missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Ping Pong Balls

Catalog No. S26599
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S26599 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S26599 Supplier Eisco™ Supplier No. PINGPONG1PK12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ping Pong Balls that can be used as a general classroom supply, in physics experiments, or as a replacement in educational kits such as trebuchet.

  • Useful as a general classroom supply, a replacement for educational kit, or in physics experiments.

Specifications

Grade K to 12
Format Kit
Product Type Ping Pong Balls
Quantity 12 Pk.
Languages English
For Use With (Application) Physics Experiments and Demonstrations
Subject Physics - Motion