Learn More
Chemglass Inc Clamp, Pinch, Stainless Steel, #35
Catalog No. CG15006
Encompass
Clamp, Pinch, Stainless Steel, #35. Made entirely of stainless steel. Part numbers CG-150-01 and CG-150-03 are supplied WITHOUT a screw type locking device. All other sizes have the screw type locking device.Note: The size #102 Stainless Steel pinch clamp is available on a special order basis.
Catalog No. CG15006 Supplier Chemglass Inc Supplier No. CG15006
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clamp, Pinch, Stainless Steel, #35. Made entirely of stainless steel. Part numbers CG-150-01 and CG-150-03 are supplied WITHOUT a screw type locking device. All other sizes have the screw type locking device.Note: The size #102 Stainless Steel pinch clamp is available on a special order basis.