missing translation for 'onlineSavingsMsg'
Learn More

Pinacyanol Iodide 90.0+%, TCI America™

Catalog No. P04361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P04361G 1 g
P04365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P04361G Supplier TCI America Supplier No. P04361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-91-4
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.39
MDL Number MFCD00011975
InChI Key QWYZFXLSWMXLDM-UHFFFAOYSA-M
Synonym carbocyanine, pinacyanole, pinacyanol iodide, sensitol red, pinacyanol, eastman kodak 622, quinaldine blue, dsstox_cid_26576, dsstox_rid_81735, dsstox_gsid_46576
PubChem CID 5709754
ChEBI CHEBI:52218
IUPAC Name 1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
SMILES [I-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 480.39
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pinacyanol Iodide
RTECSNumber VC3676870
TSCA No