missing translation for 'onlineSavingsMsg'
Learn More

Pinacolone, 97%, Thermo Scientific Chemicals

Catalog No. AAA1028922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1028922 100 g
AAA1028936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1028922 Supplier Thermo Scientific Chemicals Supplier No. A1028922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 75-97-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008846
InChI Key PJGSXYOJTGTZAV-UHFFFAOYSA-N
Synonym pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PubChem CID 6416
IUPAC Name 3,3-dimethylbutan-2-one
SMILES CC(=O)C(C)(C)C

Specifications

Melting Point °C to 53°C
Density 0.803 g/mL
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Assay Percent Range 97%
Refractive Index 1.397
Linear Formula (CH3)3CC(O)CH3
Quantity 100 g
UN Number UN1224
Merck Index 14,7440
Solubility Information Miscible with water,chloroform,ethanol,ether,acetone and carbon tetrachloride.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material Pinacolone
Show More Show Less
Hazard Category H225-H302
Hazard Statement GHS H Statement
H225-H302
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 200-920-4
RTECSNumber EL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only