missing translation for 'onlineSavingsMsg'
Learn More

Pinacolborane, 97%

Catalog No. AAL1755814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1755814 25 g
AAL1755806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1755814 Supplier Thermo Scientific Chemicals Supplier No. L1755814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25015-63-8 | C6H13BO2 | 127.98 g/mol

Pinacolborane is used as a precursor of boronic esters by hydroboration or coupling reactions. It acts as a monofunctional hydroborating agent. It is used in the synthesis of unsymmetrical biaryls through aromatic C-H borylation-cross-coupling sequence. It is involved in the preparation of 4,4,5,5-tetramethyl-2-thiophen-2-yl-[1,3,2]dioxaborolane by reacting with 2-iodo-thiophene. Further, it is used to prepare vinylboronates by palladium-catalyzed coupling with alkenyl triflates and iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolborane is used as a precursor of boronic esters by hydroboration or coupling reactions. It acts as a monofunctional hydroborating agent. It is used in the synthesis of unsymmetrical biaryls through aromatic C-H borylation-cross-coupling sequence. It is involved in the preparation of 4,4,5,5-tetramethyl-2-thiophen-2-yl-[1,3,2]dioxaborolane by reacting with 2-iodo-thiophene. Further, it is used to prepare vinylboronates by palladium-catalyzed coupling with alkenyl triflates and iodides.

Solubility
Miscible with ether, dichloromethane, tetrahydrofuran and organic solvents.

Notes
Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, water, bases and alcohols.

Chemical Identifiers

CAS 25015-63-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
MDL Number MFCD00674030
InChI Key UCFSYHMCKWNKAH-UHFFFAOYSA-N
Synonym pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n
PubChem CID 6364989
IUPAC Name 4,4,5,5-tetramethyl-1,3,2$l^{2}-dioxaborolane
SMILES CC1(C)OBOC1(C)C

Specifications

Density 0.882
Boiling Point 42°C to 43°C (50 mmHg)
Flash Point 5°C (41°F)
Refractive Index 1.397
Quantity 25 g
UN Number UN3399
Beilstein 7802871
Sensitivity Air sensitive
Solubility Information Miscible with ether,dichloromethane,tetrahydrofuran and organic solvents.
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material Pinacolborane
Show More Show Less
Hazard Category H225-H261-H315-H319-H335
Hazard Statement GHS H Statement
H225-H261-H319
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes serious eye irritation.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only