missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pimelic Acid 98.0+%, TCI America™
$104.43 - $1,087.90
Chemical Identifiers
| CAS | 111-16-0 |
|---|---|
| Molecular Formula | C7H12O4 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00004425 |
| InChI Key | WLJVNTCWHIRURA-UHFFFAOYSA-N |
| Synonym | pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f |
| PubChem CID | 385 |
| ChEBI | CHEBI:30531 |
| IUPAC Name | heptanedioic acid |
| SMILES | OC(=O)CCCCCC(O)=O |
Chemical Identifiers
| 111-16-0 | |
| 160.17 | |
| WLJVNTCWHIRURA-UHFFFAOYSA-N | |
| 385 | |
| heptanedioic acid |
| C7H12O4 | |
| MFCD00004425 | |
| pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f | |
| CHEBI:30531 | |
| OC(=O)CCCCCC(O)=O |
Specifications
| 111-16-0 | |
| White | |
| C7H12O4 | |
| 25 g | |
| WLJVNTCWHIRURA-UHFFFAOYSA-N | |
| heptanedioic acid | |
| 385 | |
| 160.17 | |
| Crystalline Powder |
| 106°C | |
| 215°C | |
| MFCD00004425 | |
| pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f | |
| OC(=O)CCCCCC(O)=O | |
| 160.17 | |
| CHEBI:30531 | |
| ≥98.0% (GC,T) | |
| Pimelic Acid |
Safety and Handling
EINECSNumber : (2)-3795
RTECSNumber : TK3677000
TSCA : Yes