missing translation for 'onlineSavingsMsg'
Learn More

Pimelic Acid 98.0+%, TCI America™

Catalog No. P043525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P043525G 25 g
P0435100G 100 g
P0435500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P043525G Supplier TCI America Supplier No. P043525G

Chemical Identifiers

CAS 111-16-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004425
InChI Key WLJVNTCWHIRURA-UHFFFAOYSA-N
Synonym pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f
PubChem CID 385
ChEBI CHEBI:30531
IUPAC Name heptanedioic acid
SMILES OC(=O)CCCCCC(O)=O

Specifications

Melting Point 106°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pimelic Acid
EINECSNumber (2)-3795
RTECSNumber TK3677000
TSCA Yes