missing translation for 'onlineSavingsMsg'
Learn More

Pimelic acid, 98%

Catalog No. AAA1849514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1849514 25 g
AAA1849522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1849514 Supplier Thermo Scientific Chemicals Supplier No. A1849514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-16-0 | C7H12O4 | 160.17 g/mol

Pimelic acid derivatives are involved in the biosynthesis of the amino acid called lysine. It is applied as a precursor to many polyesters and polyamides. It is the final member of the mnemonic used to aid recollection of the order of the first six dicarboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pimelic acid derivatives are involved in the biosynthesis of the amino acid called lysine. It is applied as a precursor to many polyesters and polyamides. It is the final member of the mnemonic used to aid recollection of the order of the first six dicarboxylic acids.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 111-16-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004425
InChI Key WLJVNTCWHIRURA-UHFFFAOYSA-N
Synonym pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f
PubChem CID 385
ChEBI CHEBI:30531
IUPAC Name heptanedioic acid
SMILES OC(=O)CCCCCC(O)=O

Specifications

Melting Point 102°C to 106°C
Density 1.329
Boiling Point 212°C (10 mmHg)
Linear Formula HO2C(CH2)5CO2H
Quantity 25 g
Beilstein 1210024
Merck Index 14,7431
Solubility Information Soluble in water. Solubility in methanol is almost transparent.
Formula Weight 160.17
Percent Purity ≥98%
Chemical Name or Material Pimelic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-840-8
RTECSNumber TK3677000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only