missing translation for 'onlineSavingsMsg'
Learn More

Pimelic Acid, 98%, Spectrum™ Chemical

Catalog No. 18612377
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612377 25 g Amber Glass Bottle
18612376 100 g Amber Glass Bottle
18612380 500 g Amber Glass Bottle
18612379 5 kg Amber Glass Bottle
18612378 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612377 Supplier Spectrum Chemical Mfg Cor Supplier No. P209725GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 111-16-0
Molecular Weight (g/mol) 160.17 g/mol

Specifications

CAS Max % 1
Color White
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Pimelic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.