missing translation for 'onlineSavingsMsg'
Learn More

Pimelic Acid, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P2097100GM

 View more versions of this product

Catalog No. 18-612-376


Only null left

Description

Description

Specifications

Specifications

Pimelic Acid
1
Amber Glass Bottle
160.17 g/mol
Reagent
111-16-0
White
100 g
0.98
Crystals or Flakes

Chemical Identifiers

111-16-0
160.17 g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.