missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pimecrolimus

Catalog No. AC464995000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464995000 500 mg
AC464991000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464995000 Supplier Thermo Scientific Chemicals Supplier No. 464995000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137071-32-0
Molecular Formula C43 H68 Cl N O11

Specifications

Color White to beige
Purity ≥96.0% by HPLC
Packaging Glass Bottle
Content And Storage Refrigerator +4°C
Shipping Condition normal conditions
Source synthetic source
Quantity 500 mg
Product Type Pimecrolimus
Molecular Weight (g/mol) 810.45
Form Powder

RUO – Research Use Only