missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134602
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P04341G 1 g
P04345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P04341G Supplier TCI America Supplier No. P04341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54-71-7
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00012722
InChI Key RNAICSBVACLLGM-GNAZCLTHSA-N
Synonym pilocarpine hydrochloride, +-pilocarpine hydrochloride, pilocarpal, pilocar, pilocar smp, almocarpine, pilomiotin, pilovisc, pilocel, pilocarpine hcl
PubChem CID 5909
IUPAC Name hydrogen (3S,4R)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one chloride
SMILES [H+].[Cl-].CC[C@H]1[C@@H](CC2=CN=CN2C)COC1=O

Specifications

Melting Point 203°C
Color White
Quantity 1 g
UN Number 1544
Formula Weight 244.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pilocarpine Hydrochloride
RTECSNumber TK1450000
TSCA Yes