Learn More
SignalChem Biotech PIP5K3 Protein 100ug
Catalog No. 502573839
Encompass
Recombinant human PIP5K3 (1493-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573839 Supplier SignalChem Biotech Supplier No. P1731G100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human PIP5K3 (1493-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.