Learn More
Acrobiosystems Recombinant Protein;Human pIgR Protein;HEK293;100UG;PIR,PIGR,Poly-Ig receptor DFS Item
Supplier:  ACROBiosystems Inc PIRH5222100UG
Recombinant Protein;Human pIgR Protein;100UG;Human pIgR, His Tag (PIR-H5222) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Arg 638 (Accession # AAI10495.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.7 kDa. The protein migrates as 85-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8000
Only null left
Explore available promotions