missing translation for 'onlineSavingsMsg'
Learn More

Pigment Red 4, TCI America™

Catalog No. p105025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P105025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P105025G Supplier TCI America Supplier No. P105025G
Only null left

Chemical Identifiers

CAS 2814-77-9
Molecular Formula C16H10ClN3O3
Molecular Weight (g/mol) 327.724
MDL Number MFCD00059095
InChI Key WJJZURXPOSMHKT-MNDPQUGUSA-N
Synonym Permanent Red 2B
PubChem CID 5720319
IUPAC Name (1Z)-1-[(2-chloro-4-nitrophenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=C(C=C(C=C3)[N+](=O)[O-])Cl

Specifications

Formula Weight 327.72
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Pigment Red 4
EINECSNumber (5)-3210
RTECSNumber QL3436000
TSCA Yes