missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pigment Orange 43 94.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 412.41 |
| Percent Purity | ≥94.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pigment Orange 43 |
Safety and Handling
| EINECSNumber | (5)-2201&(5)-3205 |
| RTECSNumber | DX1000000 |
| TSCA | Yes |