missing translation for 'onlineSavingsMsg'
Learn More

Pigment Orange 43 94.0+%, TCI America™

Catalog No. P212425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P212425G 25 g
P21245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P212425G Supplier TCI America Supplier No. P212425G
Only null left

Chemical Identifiers

CAS 4424-06-0
Molecular Formula C26H12N4O2
Molecular Weight (g/mol) 412.408
MDL Number MFCD00071829
InChI Key DGBWPZSGHAXYGK-UHFFFAOYSA-N
Synonym Vat Orange 7
PubChem CID 78141
SMILES C1=CC=C2C(=C1)N=C3N2C(=O)C4=CC=C5C6=C(C=CC3=C46)C(=O)N7C5=NC8=CC=CC=C87

Specifications

Quantity 25 g
Formula Weight 412.41
Percent Purity ≥94.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Pigment Orange 43
EINECSNumber (5)-2201&(5)-3205
RTECSNumber DX1000000
TSCA Yes