Learn More
Pigment Green 7, TCI America™
Supplier:  TCI America P066025G
Catalog No. P066025G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Pigment Green 7
C32Cl16CuN8
Phthalocyanine Green
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C3=NC4=NC(=NC5=NC(=NC6=C7C(=C([N-]6)N=C2[N-]3)C(=C(C(=C7Cl)Cl)Cl)Cl)C8=C5C(=C(C(=C8Cl)Cl)Cl)Cl)C9=C4C(=C(C(=C9Cl)Cl)Cl)Cl.[Cu+2]
6508016
Green
Crystal/Powder
1328-53-6
MFCD00053950
ABFKYPFPQRDCGM-UHFFFAOYSA-N
1127.154
1127.15
25 g
Chemical Identifiers
1328-53-6
1127.154
ABFKYPFPQRDCGM-UHFFFAOYSA-N
6508016
C32Cl16CuN8
MFCD00053950
Phthalocyanine Green
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C3=NC4=NC(=NC5=NC(=NC6=C7C(=C([N-]6)N=C2[N-]3)C(=C(C(=C7Cl)Cl)Cl)Cl)C8=C5C(=C(C(=C8Cl)Cl)Cl)Cl)C9=C4C(=C(C(=C9Cl)Cl)Cl)Cl.[Cu+2]
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
EINECSNumber : (5)-3315
TSCA : Yes