missing translation for 'onlineSavingsMsg'
Learn More

New Pig pHydrion™ INSTA-CHEK™ pH Papers

Catalog No. 189992544
Change view
Click to view available options
Type:
pHydrion™ Insta-Chek™ pH Paper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
189992544 pHydrion™ Insta-Chek™ pH Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189992544 Supplier New Pig Supplier No. PLS236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Color chart, dispenser and polyethylene case

When unknown liquids spill, use this paper for pH checks before or after

  • Instantly indicates any pH from 0 to 13 with one of 14 colors
  • Chemically reactive paper
  • Distinct, easy-to-read colors for accurate readings
  • Color chart has checkpoints at each unit pH, determined by single-color match
  • Roll dimensions (L x W): 1.5m × 12.7mm (5 ft. × 0.5 in.)

Specifications

Type pHydrion™ Insta-Chek™ pH Paper
For Use With (Application) Identifies pH level in increments of one from 0 to 13 by single-color match
Includes 14 brilliant color comparison chart with checkpoints, dispenser and polyethylene case
pH Range 0 to 13
Weight (English) 0.15 lb.
Weight (Metric) 68 g