Learn More
Biomatik Corporation Pig Interleukin 12B (IL12B) ELISA Kit, 96 tests
Catalog No. 501502935
Encompass
Pig Interleukin 12B (IL12B) ELISA Kit for detecting IL12B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 3.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28938. Gene ID: 397076. Target Synonyms: p40; IL12-B; CLMF2; NKSF2; Natural Killer Cell Stimulatory Factor 2; Cytotoxic Lymphocyte Maturation Factor 2; Interleukin 23, P40 Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501502935 Supplier Biomatik Corporation Supplier No. EKU05213
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pig Interleukin 12B (IL12B) ELISA Kit for detecting IL12B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 3.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28938. Gene ID: 397076. Target Synonyms: p40; IL12-B; CLMF2; NKSF2; Natural Killer Cell Stimulatory Factor 2; Cytotoxic Lymphocyte Maturation Factor 2; Interleukin 23, P40 Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.