Learn More
New Pig Blood, Vomit & Urine Spill Kit
Catalog No. 19041554
Change view
Click to view available options
Capacity (Metric):
11 L
Catalog No. Capacity (Metric)
19041554 11 L
1 options
Catalog No. 19041554 Supplier New Pig Supplier No. KIT5006
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Grab-and-go clean up kit that helps you stay clean, safe, and sanitary while you take care of a spill.
  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Easy-open, resealable bag protects kit contents from dirt and moisture
  • Single-use kit is faster and more efficient than dragging a large spill kit to a small cleanup; safer and more effective than using rags or paper towels
  • Includes absorbent mat pads and loose absorbent to clean up blood, vomit, urine and other bodily fluids (each kit absorbs up to 37 oz.)
  • Biohazard disposal bag helps you control germs and reduce the risk of infection
  • Disposable gloves and mini dustpan also included to aid cleanup
  • Absorbs 2.9 Gal/Cs
  • Single use kit
Specifications
Capacity (English) 2.9 gal.
Capacity (Metric) 11 L
Type Spill Kit
For Use With (Application) Cleaning up fluids