missing translation for 'onlineSavingsMsg'
Learn More

Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals

Catalog No. AAJ64277LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64277LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64277LB0 Supplier Thermo Scientific Chemicals Supplier No. J64277LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A cell-permeable p53 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 389850-21-9
Molecular Formula C15H16BrN3O3S
Molecular Weight (g/mol) 398.275
MDL Number MFCD00398180
InChI Key NXPCMOGORSWOLH-UHFFFAOYSA-N
Synonym pifithrin-alpha, p-nitro, 1-4-nitrophenyl-2-4,5,6,7-tetrahydro-2-imino-3 2h-benzothiazolyl ethanone, hbr, pifithrin-alpha, p-nitro hydrobromide, 2-2-imino-4,5,6,7-tetrahydro-1,3-benzothiazol-3-yl-1-4-nitrophenyl ethanone hydrobromide, 2-2-imino-4,5,6,7-tetrahydrobenzo d thiazol-3 2h-yl-1-4-nitrophenyl ethanone hbr, 2-2-imino-4,5,6,7-tetrahydrobenzo d thiazol-3 2h-yl-1-4-nitrophenyl ethanone hydrobromide
PubChem CID 11553038
IUPAC Name 2-(2-imino-4,5,6,7-tetrahydro-1,3-benzothiazol-3-yl)-1-(4-nitrophenyl)ethanone;hydrobromide
SMILES C1CCC2=C(C1)N(C(=N)S2)CC(=O)C3=CC=C(C=C3)[N+](=O)[O-].Br

Specifications

Color Brown
Quantity 5 mg
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in DMSO or ethanol
Formula Weight 398.3
Physical Form Solid
Chemical Name or Material Pifithrin-alpha, p-Nitro hydrobromide
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only