missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PIEZO2 Antibody Blocking Peptide

Catalog No. NB398511
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398511 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398511 Supplier R&D Systems™ Supplier No. NBP178538PEP0.1MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Blocking Peptide

Specifications

Gene ID (Entrez) 63895
Species Human
Product Line Novus
Accession Number Q9H5l5
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-78538. For further blocking peptide related protocol, click here .
Gene Alias C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
Gene Symbol PIEZO2
Quantity 0.1 mg
Research Category Alzheimers Research, Neuroscience
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Show More Show Less