Learn More
Celltreat Scientific Products Pierceable Sealing Film, Sterile
Catalog No. 5000144351
Encompass
Easily retrieve individual well samples without removing or puncturing sealing filmPre-cut flaps over each well allow easy manual or robotic pipette tip entry without the risk of adhesive residue contaminationFlaps self-seal after pipette tip is removed, ideal for short-term storage of precious samplesBoldly printed alignment circles ensure accurate film placement on standard 96 well platesEach well is individually alphanumerically labeled
Catalog No. 5000144351 Supplier Celltreat Scientific Products Supplier No. 229136
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Easily retrieve individual well samples without removing or puncturing sealing filmPre-cut flaps over each well allow easy manual or robotic pipette tip entry without the risk of adhesive residue contaminationFlaps self-seal after pipette tip is removed, ideal for short-term storage of precious samplesBoldly printed alignment circles ensure accurate film placement on standard 96 well platesEach well is individually alphanumerically labeled