missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Zinc Reversible Stain Kit

Visualize proteins in polyacrylamide gels immediately after electrophoresis and then erase the stain for subsequent transfer or band extraction.

Supplier:  Thermo Scientific™ LSG24582

Catalog No. PI24582


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce Zinc Reversible Stain Kit provides rapid and highly sensitive detection of protein in polyacrylamide gels immediately after electrophoresis as well as erasability for subsequent transfer or band extraction.

This zinc-based protein gel stain provides rapid detection of proteins after their separation by SDS-PAGE. The method is as sensitive as most silver staining techniques in that it detects protein at less than 1ng per band, yet requires less than 15 minutes to complete. The simple, two-step staining protocol results in a stained gel with an opaque milky-white background and clear protein bands. The clear bands of protein are visible when the gel is placed against a dark background.

The Pierce Zinc Reversible Stain Kit is an ideal option for in-gel protein detection preceding many downstream applications, because the zinc-stain is completely reversible. The white background is easily removed and the gel is restored to its original condition with the kit-supplied eraser solution or a Tris-glycine buffer. Afterwards, the proteins are free of any dyes or other contaminants and can be restained by another method, transferred to membrane for Western blotting or eluted from an excised gel piece.

Highlights:

  • Sensitive – detect 0.25ng of protein per band in a mini gel

  • Fast – the entire protocol is complete in 15 minutes

  • Easy – ready-to-use stain and developer solutions are supplied, and no fixing or washing steps are required

  • Gentle – no chemical modification of the proteins occurs; protein bands are visible because the background is stained, not the bands themselves

  • Reversible – completely erase the stain from the gel to easily recover proteins or proceed with other methods

Includes:

Zinc stain, developer, and eraser solutions (500mL each)

Recommended for:

Staining preceding protein recovery for antibody generation or immunological detection; Staining before excising gel pieces for protein digestion and analysis by mass spectrometry; Staining before electrophoretic transfer to membrane for Western blotting; Quick purity checks before proceeding with purification or transfer for Western blotting

Specifications

Specifications

Sufficient For: 20 mini gels (SDS-PAGE)
• Zinc Stain, 500 mL
• Zinc Developer, 500 mL
• Zinc Eraser, 500 mL

Store kit at room temperature.
Colorimetric
1.5 L
Pierce
In-Gel Detection
Proprietary Mix
Protein
Reversible Protein Gel Stain Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.