missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade

Catalog No. PI28904
Change view
Click to view available options
Quantity:
100 g
10 x 1 g
10 x 1 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28904 10 x 1 mL
PI28902 10 x 1 g
PI28903 100 g
PI28901 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI28904 Supplier Thermo Scientific™ Supplier No. LSG28904

Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

Thermo Scientific™ Pierce Trifluoroacetic Acid (TFA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.

For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.

Highlights:

  • Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
  • High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
  • Choice of formats for convenience: 1 mL ampules can prepare 1 L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
  • CAS: 76-05-1

Storage Information

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
  • Should be stored from 20° to 25°C

Order Info

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Name Note Trifluoroacetic Acid (TFA)
Quantity 10 x 1 mL
Physical Form Liquid
Percent Purity 99.5% (TFA)

For Research Use Only. Not for use in diagnostic procedures.