missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TEMED

Catalog No. PI17919
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17919 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17919 Supplier Thermo Scientific™ Supplier No. 17919
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently catalyze polyacrylamide gel polymerization in SDS-PAGE using TEMED.

Thermo Scientific™ Pierce Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization.

Additional Information and Online Ordering

TEMED is used with ammonium persulfate (APS) to catalyze acrylamide polymerization when preparing gels for electrophoresis. The chemical is a liquid at room temperature.

Highlights:

  • Chemical name: N,N,N',N'-tetramethylethane-1,2-diamine
  • Formula: C6H16N2
  • Molecular weight: 116.21
  • CAS number: 110-18-9
  • Purity: >99%
  • Refractive index: 1.417 to 1.419
  • Boiling range: 119 to 121°C

Compatible with:

Ammonium Persulfate (Part No. 17874)

Specifications

Content And Storage Store locked up in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, and separate from oxidizing materials.
Form Liquid
Quantity 25 mL
Product Type TEMED
What is the recommended storage condition for TEMED?

We recommend storing TEMED locked up in a segregated and approved area. Store in the original container protected from direct sunlight in a dry, cool and well-ventilated area and separate from oxidizing materials.

What are the Quality Specifications for each lot of TEMED?

The Certificate of Analysis lists actual test results for each lot and the minimum specification requirements:
-Purity > 99.0%
-Refractive index (nD20) between 1.417-1.419
-Boiling Range: 119-121 degrees C
-The solution must be a clear liquid, free of particulate matter

What are the characteristics of TEMED (Cat. No. 17919)?

>Chemical name: N,N,N',N'- tetramethylethane-1,2-diamine
-Formula: C6H16N2
-CAS number: 110-18-9
-Molecular weight: 116.21-Purity: ≥ 99%
-Refractive index: 1.417 to 1.419
-Boiling range: 119 to 121 degrees C
-Clear liquid at room temperature, sold as a 25 mL volume in an amber bottle

What is Thermo Scientific Tetramethylethylenediamine (TEMED)?

Thermo Scientific Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is used with ammonium persulfate (APS) to catalyze acrylamide polymerization when preparing gels for electrophoresis.

Do you carry ammonium persulfate and TEMED?

Yes, details below:

- Ammonium Persulfate: Cat. No. 17874
- TEMED: Cat. No. 15524010 and Cat. No. 17919

For Research Use Only. Not for use in diagnostic procedures.