Learn More
Thermo Scientific™ Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
For picogram-level detection and Western blot development less than 1 hour after gel transfer.
Supplier: Thermo Scientific™ 35061
Description
Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol that allows picogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.
These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.Highlights:
Fast – picogram levels of detection in about one hour
Convenient – no expensive hardware or vacuum required; no clogging issues
Simple – optimized protocol makes Western blot analysis easier than ever
Economical – no additional consumables or extra equipment to purchase
Stable – all kit components are stable for one year stored at 4°C
Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody
Compatible with:
Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer
Specifications
| Western Blot Kit | |
| Liquid | |
| Nitrocellulose, PVDF | |
| 800 mL | |
| Horseradish Peroxidase | |
| Picogram-level | |
| Sufficient For: 20 mini blots probed with rabbit primary antibodies • Fast Western Antibody Diluent, 400 mL • Fast Western 10X Wash Buffer, 200 mL • Fast Western Rabbit Optimized HRP Reagent, Pico, 20 mL • SuperSignal West Pico Luminol/Enhancer Solution, 100 mL • SuperSignal West Stable Peroxide Solution, 100 mL Store kit at 4°C. |
| Chemiluminescence | |
| Western Blot | |
| Pierce, SuperSignal | |
| HRP | |
| Rabbit | |
| 20 Mini Blots |
For Research Use Only. Not for use in diagnostic procedures.