missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated High Capacity Plates

Catalog No. PI15505
Change view
Click to view available options
Color:
Black
Clear
White
No. of Wells:
384
96
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Format Detection Method
PI15505 White 384 Standard Chemiluminescence
PI15503 Black 96 Standard Fluorescence
PI15506 Black 384 Standard Fluorescence
PI15501 Clear 96 Strip Plate Colorimetric
PI15500 Clear 96 Standard Colorimetric
PI15504 Clear 384 Standard Colorimetric
PI15502 White 96 Standard Chemiluminescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. PI15505 Supplier Thermo Scientific™ Supplier No. LSG15505

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce Streptavidin High Binding Capacity (HBC) Coated Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors, and coating volumes.

Thermo Scientific Pierce Streptavidin-Coated High-Capacity Plates are designed for the binding of biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin-coated plates are widely used for applications such as ELISAs and other assay protocols. These biotin-binding plates come with different capacities and sensitivities for maximum flexibility. For example, the high-binding-capacity plates (HBC) can be used for anti-therapeutic/anti-drug antibodies (ADA) screening for immunogenicity research and biologics development (1,2). Both regular and HBC plates have been used in aptamer assays for clinical and industrial needs, along with neurobiology research for therapeutics and prevention, amongst other applications (3–5).

Features of Streptavidin-Coated High-Capacity Plates include:

  • Unique plate-coating technology—results in high loading and binding capacity
  • Streptavidin-coated option—plates have up to five times the binding capacity of typical plates
  • Broader dynamic range—extends the quantitative range so there is less need for dilutions
  • Save time—pre-blocked plates to reduce the number of assay steps
  • Flexible assay formats—coated plates offered in 8-well strips to reduce waste

The proprietary plate-coating technology used to manufacture these Pierce coated plates yields high-capacity plates with a wider detection limit than regular-binding-capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

References

  1. J Immunol Methods. 2016 Apr;431:45-51
  2. J Immunol Methods. 2011 Feb 28;365(1-2):38-49
  3. Cancers (Basel). 2022 Oct 25;14(21):5222
  4. J Microbiol Biotechnol. 2013 Jun 28;23(6):878-84
  5. Acta Neuropathol Commun. 2018 May 31;6(1):43

Specifications

Binding Property High Capacity, Affinity
Format Standard
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.
Detection Method Chemiluminescence
Product Line Pierce
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Surface Treatment Streptavidin
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less
How has Streptavidin been linked to the Pierce Streptavidin Coated High Capacity Plates (Cat. No. 15500)?

The linking of streptavidin has been performed by passive binding. This means the molecule has been attached to the plates just by simple incubation (see also https://www.thermofisher.com/de/en/home/life-science/protein-biology/protein-assays-analysis/elisa/elisa-microplates-plasticware/adsorption-immunoassay-plates.html).

What is the shelf life of Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well?

Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.